Skip to Content
Merck
All Photos(1)

Key Documents

900308

Sigma-Aldrich

CdS/ZnS core-shell type quantum dots

lyophilized, fluorescence λem 425 nm, solid

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

lyophilized
powder
solid

λmax

410 nm

fluorescence

λem 425 nm
λem 425 nm±10 nm, quantum yield >50%

functional group

carboxylic acid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

CdS/ZnS or CdSe/ZnS quantum dots (QDs) with carboxylic acid in lyophilized solid form can be readily reconstituted in DI water or DMSO to any desired concentration and are very stable in most buffer solutions with a pH range of 5-10. These QDs possess a zeta potential in the -30 mV to -50 mV range and can easily be reacted with amine containing molecules in DMSO or water. These QDs find applications in bioimaging and biosensing.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single chain epidermal growth factor receptor antibody conjugated nanoparticles for in vivo tumor targeting and imaging.
Yang L, et al.
Small, 5, 235-243 (2009)
Quantum dot-based, quantitative, and multiplexed assay for tissue staining.
Xu H, et al.
ACS Applied Materials & Interfaces, 5, 2901-2907 (2013)
Zengyan Wei et al.
Soft matter, 8(26), 6871-6875 (2012-09-18)
Genetically-engineered collagen peptides were assembled into freestanding films when QDs are co-assembled as joints between collagen domains. These peptide based films show excellent mechanical properties with Young's modulus of ~20 GPa, much larger than most of multi-composite polymer films and

Articles

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service