Skip to Content
Merck
All Photos(1)

Key Documents

900746

Sigma-Aldrich

Perovskite quantum dots

oleic acid and oleylamine coated, fluorescence λem 510 nm, 10 mg/mL in toluene

Synonym(s):

Cadmium free QDs, Fluorescent nanocrystals, Perovskite nanocrystals, QDs

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€370.00

€370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€370.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

€370.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

concentration

10 mg/mL in toluene

fluorescence

λem 510 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For Perovskite quantum dots:
Perovskite quantum dots (QDs) of common formula CsPbX3 (X = Cl, Br, I) possess high photoluminescence efficiency and narrow emission and emit in the visible spectral regime. Perovskite QDs are cadmium free and the aforementioned properties render them suitable for applications in light emitting diodes (LEDs), lasers, liquid crystal displays (LCDs) etc.[1][2]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loredana Protesescu et al.
Nano letters, 15(6), 3692-3696 (2015-01-31)
Metal halides perovskites, such as hybrid organic-inorganic CH3NH3PbI3, are newcomer optoelectronic materials that have attracted enormous attention as solution-deposited absorbing layers in solar cells with power conversion efficiencies reaching 20%. Herein we demonstrate a new avenue for halide perovskites by
CsPbX3 Quantum Dots for Lighting and Displays: Room-Temperature Synthesis, Photoluminescence Superiorities, Underlying Origins and White Light-Emitting Diodes.
Li X, et al.
Advances in Functional Materials, 26(16), 2435-2445 (2016)

Articles

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

See All

Questions

  1. What is the size of these PQDs.

    1 answer
    1. The size of this product is approximately 10 nanometers.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service