Skip to Content
Merck
All Photos(1)

Key Documents

86911

Sigma-Aldrich

Tetrabutylammonium heptadecafluorooctanesulfonate

≥95.0% (H-NMR)

Synonym(s):

Heptadecafluorooctanesulfonic acid tetrabutylammonium salt, Perfluorooctanesulfonic acid tetrabutylammonium salt, Tetrabutylammonium perfluorooctanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[CF3(CF2)7SO3]
CAS Number:
Molecular Weight:
741.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (H-NMR)

form

liquid

functional group

amine
fluoro
sulfonic acid

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C16H36N.C8HF17O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28/h5-16H2,1-4H3;(H,26,27,28)/q+1;/p-1

InChI key

MUOQTHSUZGSHGW-UHFFFAOYSA-M

Application

Tetrabutylammonium heptadecafluorooctanesulfonate [NBu4][(PFOc)SO3] can be used as:
  • A model ionic liquid for the possible replacement of perfluorocarbons, which are used in oxygen therapeutics.
  • A supporting electrolyte in the electropolymerization of 3,4-ethylenedioxythiophene (EDOT) derivatives to obtain superhydrophobic surfaces.

Other Notes

Ionic liquid

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.M. Pomaville et al.
Journal of Chromatography A, 499, 749-749 (1990)
T.O. Kollie et al.
Chromatographia, 33, 551-551 (1992)
(Liquid+ liquid) equilibria of perfluorocarbons with fluorinated ionic liquids
Martinho, S., et al.
The Journal of Chemical Thermodynamics, 64, 71-79 (2013)
Versatile superhydrophobic surfaces from a bioinspired approach
Wolfs M, et al.
Macromolecules, 44(23), 9286-9294 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service