Skip to Content
Merck
All Photos(1)

Key Documents

924407

Sigma-Aldrich

1H,1H,2H,2H-Tridecafluorooctane-1-sulphonic acid

Synonym(s):

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanesulfonic acid, 1H,1H′,2H,2H′-Perfluorooctane sulfonic acid, 1H,1H,2H,2H-Perfluoro-1-octanesulfonic acid, 6:2 Fluorotelomer sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€129.00

About This Item

Empirical Formula (Hill Notation):
C8H5F13O3S
CAS Number:
Molecular Weight:
428.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

€129.00


Check Cart for Availability

Request a Bulk Order

storage temp.

2-8°C

Quality Level

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[S](=O)(=O)O

InChI

1S/C8H5F13O3S/c9-3(10,1-2-25(22,23)24)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1-2H2,(H,22,23,24)

InChI key

VIONGDJUYAYOPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service