Skip to Content
Merck
All Photos(1)

Key Documents

859052

Sigma-Aldrich

N-Hippuryl-His-Leu hydrate

99%, for peptide synthesis

Synonym(s):

N-Benzoyl-Gly-His-Leu, N-Hippuryl-L-histidyl-L-leucine hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€96.20

€96.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€96.20

About This Item

Empirical Formula (Hill Notation):
C21H27N5O5 · xH2O
CAS Number:
Molecular Weight:
429.47 (anhydrous basis)
Beilstein:
5664178
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€96.20


Check Cart for Availability

Request a Bulk Order

Product Name

N-Hippuryl-His-Leu hydrate, 99%

Quality Level

Assay

99%

form

powder

optical activity

[α]22/D −41°, c = 1 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

108-110 °C (dec.)

application(s)

peptide synthesis

SMILES string

O.CC(C)C[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CNC(=O)c2ccccc2)C(O)=O

InChI

1S/C21H27N5O5.H2O/c1-13(2)8-17(21(30)31)26-20(29)16(9-15-10-22-12-24-15)25-18(27)11-23-19(28)14-6-4-3-5-7-14;/h3-7,10,12-13,16-17H,8-9,11H2,1-2H3,(H,22,24)(H,23,28)(H,25,27)(H,26,29)(H,30,31);1H2/t16-,17-;/m0./s1

InChI key

NWOJMNXIJBZMPK-QJHJCNPRSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

NBz-Gly-His-Leu

Substrates

Substrate for angiotensin converting enzyme.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service