Skip to Content
Merck
All Photos(2)

Key Documents

810002

Sigma-Aldrich

Phenylcyclohexane

battery grade, ≥99%, acid < 200 ppm, H2O < 100 ppm

Synonym(s):

Cyclohexylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€258.00

€258.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€258.00

About This Item

Linear Formula:
C6H5C6H11
CAS Number:
Molecular Weight:
160.26
Beilstein:
1906803
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

€258.00


Check Cart for Availability

Request a Bulk Order

grade

battery grade

Quality Level

Assay

≥99%

form

liquid

impurities

≤100 ppm H2O
≤200 ppm acid

refractive index

n20/D 1.526 (lit.)

bp

239-240 °C (lit.)

mp

4-7 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

C1CCC(CC1)c2ccccc2

InChI

1S/C12H16/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1,3-4,7-8,12H,2,5-6,9-10H2

InChI key

IGARGHRYKHJQSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These additives have low water content (less than 100 ppm), please handle under inert and moisture free enviornment (glove box). Keep containers tightly closed. Keep away from heat and ignition sources. Store in a cool and dry place. Avoid storing together with oxidizers.

Application

The presence of electrolyte additives enhances the performance of lithium ion batteries by improving their thermal stability and ionic conductivity. Additives also reduce the flammability of electrolytes and provide overcharge protection. Phenylcyclohexane finds application as an additive in lithium ion batteries.[1][2]

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu M, et al.
Journal of Power Sources, 184, 427-427 (2008)
Zhang S.
Journal of Power Sources, 162, 1379-1379 (2006)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Due to the adverse impact of the continued use of fossil fuels on the earth’s environment and climate, researchers have been asked to develop new approaches for producing power using renewable sources like wind and solar energy

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service