Skip to Content
Merck
All Photos(2)

Key Documents

809985

Sigma-Aldrich

1,3-Propanesultone

battery grade, ≥99%, acid <200 ppm, H2O <100 ppm

Synonym(s):

1,2-Oxathiolane 2,2-dioxide, 3-Hydroxypropanesulfonic acid γ-sultone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€266.00
500 G
€1,010.00

€266.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€266.00
500 G
€1,010.00

About This Item

Empirical Formula (Hill Notation):
C3H6O3S
CAS Number:
Molecular Weight:
122.14
Beilstein:
109782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

€266.00


Check Cart for Availability

Request a Bulk Order

grade

battery grade

Quality Level

description

Acid content: <10 ppm

Assay

≥99%

form

powder

impurities

≤100 ppm H2O
≤200 ppm acid

bp

180 °C/30 mmHg (lit.)

mp

30-33 °C (lit.)

density

1.392 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=S1(OCCC1)=O

InChI

1S/C3H6O3S/c4-7(5)3-1-2-6-7/h1-3H2

InChI key

FSSPGSAQUIYDCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The presence of electrolyte additives enhances the performance of lithium ion batteries by improving their thermal stability and ionic conductivity. Additives also reduce the flammability of electrolytes and provide overcharge protection. 1,3-Propanesultone finds application as an additive in lithium ion batteries.[1][2]

Other Notes

These additives have low water content (<100 ppm). Please handle under inert and moisture free environment (glove box). Keep containers tightly closed. Keep away from heat and ignition sources. Store in a cool and dry place. Avoid storing together with oxidizers. Application as an additive in lithium ion batteries.

Legal Information

Product of MU Ionic Solutions Corp

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yim T, et al.
Royal Society of Chemistry Advances, 4, 19172-19172 (2014)
Zhang S.
Journal of Power Sources, 162, 1379-1379 (2006)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Due to the adverse impact of the continued use of fossil fuels on the earth’s environment and climate, researchers have been asked to develop new approaches for producing power using renewable sources like wind and solar energy

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service