Skip to Content
Merck
All Photos(1)

Documents

808334

Sigma-Aldrich

Ethylbenzene-13C8

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CH213C6H5
Molecular Weight:
114.11
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+8

SMILES string

[13CH3][13CH2][13C]1=[13CH][13CH]=[13CH][13CH]=[13CH]1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1

InChI key

YNQLUTRBYVCPMQ-LLXNCONNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service