Skip to Content
Merck
All Photos(3)

Key Documents

808245

Sigma-Aldrich

10,10′-dibromo-9,9′-bianthracene

Synonym(s):

10,10′-dibromo-9,9′-bianthryl, 10-bromo-9-(10-bromoanthracen-9-yl)anthracene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€201.00
5 G
€765.00

€201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€201.00
5 G
€765.00

About This Item

Empirical Formula (Hill Notation):
C28H16Br2
CAS Number:
Molecular Weight:
512.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€201.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

mp

348-352 °C

SMILES string

BrC1=C2C(C=CC=C2)=C(C3=C(C=CC=C4)C4=C(Br)C5=C3C=CC=C5)C6=CC=CC=C61

InChI

1S/C28H16Br2/c29-27-21-13-5-1-9-17(21)25(18-10-2-6-14-22(18)27)26-19-11-3-7-15-23(19)28(30)24-16-8-4-12-20(24)26/h1-16H

InChI key

NPNNLGXEAGTSRN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface-Assisted Reactions toward Formation of Graphene Nanoribbons on Au(110) Surface.
Lorenzo M, et al.
The Journal of Physical Chemistry C, 119(5), 2427-2437 (2015)
Yen-Chia Chen et al.
Nature nanotechnology, 10(2), 156-160 (2015-01-13)
Bandgap engineering is used to create semiconductor heterostructure devices that perform processes such as resonant tunnelling and solar energy conversion. However, the performance of such devices degrades as their size is reduced. Graphene-based molecular electronics has emerged as a candidate
Akimitsu Narita et al.
Chemical record (New York, N.Y.), 15(1), 295-309 (2014-11-22)
In this article, we describe our chemical approach, developed over the course of a decade, towards the bottom-up synthesis of structurally well-defined graphene nanoribbons (GNRs). GNR synthesis can be achieved through two different methods, one being a solution-phase process based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service