Skip to Content
Merck
All Photos(2)

Key Documents

806412

Sigma-Aldrich

2,6-Dimethylanilino(oxo)acetic acid

Synonym(s):

[(2,6-Dimethylphenyl)amino](oxo)acetic acid, [(2,6-Dimethylphenyl)carbamoyl]formic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€85.90
1 G
€135.00

€85.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€85.90
1 G
€135.00

About This Item

Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€85.90


Check Cart for Availability

Request a Bulk Order

Assay

95%

Quality Level

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

O=C(C(O)=O)NC1=C(C)C=CC=C1C

InChI

1S/C10H11NO3/c1-6-4-3-5-7(2)8(6)11-9(12)10(13)14/h3-5H,1-2H3,(H,11,12)(H,13,14)

InChI key

JFTAPSBIODPQJN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

KitAlysis™ Cu C-N (Buchwald-Hartwig) cross-coupling high-throughput screening kit. Detailed Set-Up User Guide and a downloadable excel file for calculations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service