Skip to Content
Merck
All Photos(1)

Key Documents

804533

Sigma-Aldrich

2,2′-Bipyrazine

Synonym(s):

2,2′-Bipyrazinyl, 2,2′-Dipyrazinyl

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€224.00
1 G
€502.00

€224.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€224.00
1 G
€502.00

About This Item

Empirical Formula (Hill Notation):
C8H6N4
CAS Number:
Molecular Weight:
158.16
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€224.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

Quality Level

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

185-188 °C

SMILES string

n1c(cncc1)c2nccnc2

InChI

1S/C8H6N4/c1-3-11-7(5-9-1)8-6-10-2-4-12-8/h1-6H

InChI key

DFXNVSIALRDJHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Bipyrazine (bpz) can be used as a ligand to synthesize strongly oxidizing photocatalysts. Ruthenium-based homoleptic tris(bipyrazyl) complex [(Ru(bpz)32+] derived from 2,2′-bipyrazine ligand can be used as an effective photocatalyst for oxidatively induced organic transformations. For instance, [(Ru(bpz)32+] is capable of promoting radical cation mediated Diels–Alder cycloadditions, radical thiol-ene couplings, photooxygenation reactions, and [3+2] cycloaddition reactions.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visible Light Photocatalysis of Radical Cation Diels-Alder Cycloadditions: Preparation of Tris (2, 2?-bipyrazyl) Ruthenium (II) Bis (tetrakis (3, 5-bis (trifluoromethyl) phenyl) borate)
Lies SD, et al.
Organic Syntheses, 93, 178-199 (2003)
An improved procedure for the preparation of Ru (bpz) 3 (PF6) 2 via a high-yielding synthesis of 2, 2?-bipyrazine
Schultz DM, et al.
Beilstein Journal of Organic Chemistry, 11(1), 61-65 (2015)

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service