Skip to Content
Merck
All Photos(3)

Key Documents

P56003

Sigma-Aldrich

Pyrazine

≥99%

Synonym(s):

1,4-Diazabenzene, 1,4-Diazine, Paradiazine, Piazine, p-Diazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€29.40
10 G
€42.10
25 G
€72.20

€29.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€29.40
10 G
€42.10
25 G
€72.20

About This Item

Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
Beilstein:
103905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€29.40


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

crystals

bp

115-116 °C (lit.)

mp

50-56 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

SMILES string

c1cnccn1

InChI

1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H

InChI key

KYQCOXFCLRTKLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrazine is a 1,4-diazine and an electron-deficient N-heteroarene found in several natural products, APIs, agrochemicals, and functional materials.[1]

Application

Pyrazine can be used:
  • As a bridging ligand in the synthesis of Cu(II) linear chain coordination complexes.[2]
  • To prepare Cu(I) derived coordination polymers, which are used in the synthesis of metal-organic frameworks (MOFs).[3]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrazine-bridged Cu (ii) chains: diaquabis (n-methyl-2-pyridone) copper (ii) perchlorate complexes
Kirkman-Davis E, et al.
Dalton Transactions, 49(39), 13693-13703 (2020)
Microwave assisted hydrothermal synthesis of a novel CuI-sulfate-pyrazine MOF
Amo-Ochoa P, et al.
Inorganic Chemistry Communications, 10(8), 921-924 (2007)
Direct C-H photoarylation of diazines using aryldiazonium salts and visible-light
Silva RC, et al.
Royal Society of Chemistry Advances, 10(52), 31115-31122 (2020)
Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method
Nicole Favreau-Farhadi et al.
Journal of food science, 80(10), C2140-C2146 (2015-09-27)
Rosmarinic acid and Epigallocatechin gallate concentrations were studied as natural inhibitors of Maillard browning in glucose/glycine model systems, and in bakery rolls and applesauce. The concentrations of the inhibitors were varied to determine the highest level of inhibition without a

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service