![2-Bromo-1,10-phenanthroline](/deepweb/assets/sigmaaldrich/product/structures/329/917/de81954e-068e-47be-b45b-86d792e9a69d/640/de81954e-068e-47be-b45b-86d792e9a69d.png)
2-Bromo-1,10-phenanthroline
![2-Bromo-1,10-phenanthroline](/deepweb/assets/sigmaaldrich/product/structures/329/917/de81954e-068e-47be-b45b-86d792e9a69d/640/de81954e-068e-47be-b45b-86d792e9a69d.png)
Empirical Formula (Hill Notation):
C12H7BrN2
CAS Number:
Molecular Weight:
259.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing