Skip to Content
Merck
All Photos(2)

Key Documents

802182

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)

lactide:glycolide (50:50), ester terminated, average Mw 100,000

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€387.00

€387.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€387.00

About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€387.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

feed ratio

lactide:glycolide (50:50)

mol wt

average Mw 100,000

composition

lactide:glycolide (50:50)

storage temp.

2-8°C

SMILES string

OCC(O)=O.CC(O)C(O)=O

InChI key

XBBVURRQGJPTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biodegradable polymer for tissue engineering and drug delivery

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Physical form

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service