Skip to Content
Merck
All Photos(2)

Key Documents

802123

Sigma-Aldrich

6,6′-Dibromoisoindigo

Synonym(s):

6-Bromo-3-(6-bromo-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-2H-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8Br2N2O2
CAS Number:
Molecular Weight:
420.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

98%

Quality Level

form

powder

mp

>300 °C

SMILES string

O=C1NC2=C(C=CC(Br)=C2)/C1=C3C(NC4=C\3C=CC(Br)=C4)=O

InChI

1S/C16H8Br2N2O2/c17-7-1-3-9-11(5-7)19-15(21)13(9)14-10-4-2-8(18)6-12(10)20-16(14)22/h1-6H,(H,19,21)(H,20,22)/b14-13+

InChI key

TWTPKJYUWWAHQJ-BUHFOSPRSA-N

Application

Electron accepting building block for the preparation of electroactive materials for organic electronics.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Lei et al.
Journal of the American Chemical Society, 133(16), 6099-6101 (2011-04-07)
Two conjugated polymers, IIDDT and IIDT, based on an isoindigo core were developed for organic field-effect transisitors. Investigation of their field-effect performance indicated that IIDDT exhibited air-stable mobility up to 0.79 cm(2) V(-1) s(-1), which is quite high among polymer
Ergang Wang et al.
Journal of the American Chemical Society, 133(36), 14244-14247 (2011-08-19)
A new, low-band-gap alternating copolymer consisting of terthiophene and isoindigo has been designed and synthesized. Solar cells based on this polymer and PC(71)BM show a power conversion efficiency of 6.3%, which is a record for polymer solar cells based on
Jianguo Mei et al.
Organic letters, 12(4), 660-663 (2010-01-27)
Isoindigo, as a new electron acceptor unit for organic electronic materials, was integrated into two low-energy gap oligothiophenes. Optical and electrochemical studies of the newly synthesized oligomers demonstrate broad absorption through the visible spectrum, along with appropriate energy levels, as
Yi Ren et al.
ACS applied materials & interfaces, 6(16), 14533-14542 (2014-08-05)
We designed and synthesized a series of isoindigo-based derivatives to investigate how chemical structure modification at both the 6,6'- and 5,5'-positions of the core with electron-rich and electron-poor moieties affect photophysical and redox properties as well as their solid-state organization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service