Skip to Content
Merck
All Photos(3)

Documents

693847

Sigma-Aldrich

4,7-Dibromobenzo[c]-1,2,5-thiadiazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Br2N2S
CAS Number:
Molecular Weight:
293.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

184-185 °C

SMILES string

Brc1ccc(Br)c2nsnc12

InChI

1S/C6H2Br2N2S/c7-3-1-2-4(8)6-5(3)9-11-10-6/h1-2H

InChI key

FEOWHLLJXAECMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monomer for synthesis of light-emitting and conducting polymers for organic electronics.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 61, 10975-10975 (2005)
Zi Jun Wang et al.
ChemSusChem, 8(20), 3459-3464 (2015-09-10)
Metal-free visible-light photocatalysts offer a clean, sustainable solution to many pressing environmental issues. Herein, we present a molecular design strategy to fine-tune the valence and conduction band levels of a series of conjugated microporous polymer networks based on poly(benzochalcogenadiazole) for

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service