Skip to Content
Merck
All Photos(1)

Documents

797766

Sigma-Aldrich

Graphene nanoribbons

greener alternative

alkyl functionalized

Synonym(s):

GNRs, nano-graphene ribbons

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
Molecular Weight:
12.01
EC Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥85% carbon basis (TGA)

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

L × W

2-15 μm × 40-250 nm

surface area

38 m2/g

density

1.5763 g/mL (He gas method)

bulk density

0.0953 g/mL (Mercury Porosimetry)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

  • Composites.
  • Conductive inks.
  • Electrodes for LiB.
  • Energy storage & harvesting applications.
  • Bio-medical applications.

Preparation Note

Preferred dispersing organic solvents: pyrrolidones and chlorinated solvents, also dispersible in acetone, ethyl acetate, dimethyl formamide, ethylene glycol, or 1,2-dimethoxyethane at concentration ≤0.1 mg/mL after bath sonication for 15 min with stability for a few hours.
  • Less preferred dispersing organic solvents: cyclohexanone and γ-butyrolactone.
  • Aqueous dispersions are possible at 0.1 mg/mL with triton-X-100, sodium cholate and deoxycholate and cellulose-based surfactants.

Analysis Note

Raman Spectroscopic Data:
ID/IG = 1.84±0.25
I2D/IG = 2.12±0.32
2D FWHM = 41 cm-1

Legal Information

Graphene Nanoribbon Composites and Methods of Making the Same, WO/2012/112435 A1.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Throughput Preparation of Large Area Transparent Electrodes Using Non-Functionalized Graphene Naroribbons.
Zhu Y, et al.
Chemistry of Materials, 23, 935-939 (2011)
Dmitry V Kosynkin et al.
ACS nano, 5(2), 968-974 (2011-01-06)
Here we demonstrate that graphene nanoribbons (GNRs) free of oxidized surfaces can be prepared in large batches and 100% yield by splitting multiwalled carbon nanotubes (MWCNTs) with potassium vapor. If desired, exfoliation is attainable in a subsequent step using chlorosulfonic
Lei Li et al.
ACS applied materials & interfaces, 5(14), 6622-6627 (2013-06-25)
A facile and cost-effective approach to the fabrication of a nanocomposite material of polyaniline (PANI) and graphene nanoribbons (GNRs) has been developed. The morphology of the composite was characterized by scanning electron microscopy, transmission electron microscopy, X-ray photoelectron microscopy, and
Bostjan Genorio et al.
ACS nano, 6(5), 4231-4240 (2012-04-04)
A cost-effective and potentially industrially scalable, in situ functionalization procedure for preparation of soluble graphene nanoribbon (GNRs) from commercially available carbon nanotubes is presented. The physical characteristics of the functionalized product were determined using SEM, evolved gas analysis, X-ray diffraction
Changsheng Xiang et al.
ACS nano, 7(11), 10380-10386 (2013-10-10)
A thermoplastic polyurethane (TPU) composite film containing hexadecyl-functionalized low-defect graphene nanoribbons (HD-GNRs) was produced by solution casting. The HD-GNRs were well distributed within the polyurethane matrix, leading to phase separation of the TPU. Nitrogen gas effective diffusivity of TPU was

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service