Skip to Content
Merck
All Photos(2)

Key Documents

473642

Sigma-Aldrich

Glycidyl hexadecyl ether

technical grade

Synonym(s):

2,3-Epoxypropyl hexadecyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€99.10

€99.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€99.10

About This Item

Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€99.10


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

form

solid

mp

28-31 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCOCC1CO1

InChI

1S/C19H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-17-19-18-21-19/h19H,2-18H2,1H3

InChI key

YZUMRMCHAJVDRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lei Zou et al.
Journal of biomedical nanotechnology, 14(10), 1785-1795 (2018-07-26)
Convenient methods for the preparation of gene delivery platforms based on branched low molecular weight polyethylenimine (PEI) were described. Firstly, PEI lipids, with a low molecular weight PEI headgroup and hexadecyl chain tail group, were prepared through a highly efficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service