Skip to Content
Merck
All Photos(1)

Key Documents

797499

Sigma-Aldrich

1-Benzyl-6-tosyl-1,6-diazaspiro[3.3]heptan-3-ol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O3S
CAS Number:
Molecular Weight:
358.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

functional group

hydroxyl
phenyl
sulfonamide

storage temp.

2-8°C

SMILES string

OC1CN(CC2=CC=CC=C2)C13CN(S(C4=CC=C(C)C=C4)(=O)=O)C3

InChI

1S/C19H22N2O3S/c1-15-7-9-17(10-8-15)25(23,24)21-13-19(14-21)18(22)12-20(19)11-16-5-3-2-4-6-16/h2-10,18,22H,11-14H2,1H3

InChI key

LGWSZRHTXSQOOE-UHFFFAOYSA-N

Application

Spirocyclic building block is part of a series of advanced angular[3.3]heptanes developed by Carreira and coworkers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service