Skip to Content
Merck
All Photos(1)

Key Documents

795917

Sigma-Aldrich

2-Keto-3-(methyl-13C,d1)-butyric-3,4,4,4-d4 acid sodium salt

98 atom % D, 99 atom % 13C, 97% (CP)

Synonym(s):

α-Keto-isovaleric acid-13C,3,4,4,4-d4-(methyl-d1) sodium, α-Ketoisovaleric acid-13C,3,4,4,4-d4-(methyl-d1) sodium salt, 2-Ketoisovaleric acid-13C,3,4,4,4-d4-(methyl-d1) sodium salt, 3-Methyl-13C,d1-2-oxobutanoic-3,4,4,4-d4 acid sodium salt, Sodium α-ketoisovalerate-13C,d5 sodium

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€2,360.00

€2,360.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 MG
€2,360.00

About This Item

Empirical Formula (Hill Notation):
CD3CD13CDH2COCO2Na
Molecular Weight:
144.12
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€2,360.00


Estimated to ship on29 May 2025


isotopic purity

99 atom % 13C
98 atom % D

Quality Level

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+6

SMILES string

O=C(O[Na])C(C([2H])([13CH2][2H])C([2H])([2H])[2H])=O

InChI

1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1D3,2+1D,3D;/t3-;/m0./s1

InChI key

WIQBZDCJCRFGKA-YFKZDALSSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service