Skip to Content
Merck
All Photos(1)

Key Documents

571334

Sigma-Aldrich

2-Keto-3-(methyl-13C)-butyric-4-13C acid sodium salt

99 atom % 13C, 97% (CP)

Synonym(s):

α-Keto-3-(methyl-13C)-butyric-4-13C acid sodium, α-Ketoisovaleric acid-13C2 sodium salt, 2-Ketoisovaleric acid-13C2 sodium salt, 3-Methyl-13C-oxobutanoic-4-13C acid sodium salt, Sodium α-ketoisovalerate-3-(methyl-13C),4-13C

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€392.00
500 MG
€836.00
1 G
€1,800.00

€392.00


Estimated to ship on05 May 2025


Request more information

Select a Size

Change View
100 MG
€392.00
500 MG
€836.00
1 G
€1,800.00

About This Item

Linear Formula:
13CH3CH(13CH3)COCO2Na
CAS Number:
Molecular Weight:
140.08
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€392.00


Estimated to ship on05 May 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

227-230 °C (lit.)

mass shift

M+2

SMILES string

[Na+].[13CH3]C([13CH3])C(=O)C([O-])=O

InChI

1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1+1,2+1;

InChI key

WIQBZDCJCRFGKA-AWQJXPNKSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service