Skip to Content
Merck
All Photos(1)

Key Documents

793388

Sigma-Aldrich

Aluminum-doped zinc oxide nanoparticle ink

Synonym(s):

AZO dispersion, AZO ink, Al-doped ZnO ink, Al:ZnO ink, Aluminum doped zinc oxide nanoparticle dispersion, Aluminum-doped zinc oxide suspension, Avantama N-10X, Nanograde N-10X

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO/AlO3
UNSPSC Code:
12352103

form

dispersion (nanoparticle)

concentration

2.5 wt. % (crystalline Al-doped ZnO (98 wt% ZnO; 2 wt% Al2O3) in 2-propanol)

color

translucent yellowish

particle size

<50 nm (BET)

density

0.8016 g/mL at 25 °C

Application

This Al-doped ZnO nanoparticle ink is for slot-dye, spin-coating and doctor blading for the use as electron transport layer in printed electronics.
This Al-doped ZnO nanoparticle ink is universally applicable in normal and inverted architecture with performance comparable to evaporated calcium.
Annealing temperature <100°C

Other Notes

Prior to application: Ultrasonicate and (optionally) filter through 0.45 μm PTFE filter

Legal Information

Product of Avantama Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A universal method to form the equivalent ohmic contact for efficient solution-processed organic tandem solar cells
Journal of Materials Chemistry, 2, 14896?14902-14896?14902 (2014)
Flexible organic tandem solar modules with 6% efficiency: combining roll-to-roll compatible processing with high geometric fill factors
Energy & Environmental Science, 7, 3284?3290-3284?3290 (2014)
Chun-Chao Chen et al.
Advanced materials (Deerfield Beach, Fla.), 26(32), 5670-5677 (2014-07-22)
Tandem solar cells have the potential to improve photon conversion efficiencies (PCEs) beyond the limits of single-junction devices. In this study, a triple-junction tandem design is demonstrated by employing three distinct organic donor materials having bandgap energies ranging from 1.4
Interface Design to Improve the Performance and Stability of Solution-Processed Small-Molecule Conventional Solar Cells
Advanced Energy Materials (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service