Skip to Content
Merck
All Photos(2)

Key Documents

921564

Sigma-Aldrich

Aluminum doped zinc oxide nanoparticle ink

greener alternative

2.5 wt. %, viscosity 3.5 cP , work function 3.9 eVvariation )

Synonym(s):

Al:ZnO ink, Al:ZnO nanoparticle dispersion, Ink for OLED, Ink for charge selective layers in printed electronics, OPV, PV, Photodetectors, QLED

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€112.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€112.00

About This Item

Linear Formula:
Al:ZnO
CAS Number:
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.23

€112.00


Check Cart for Availability

Request a Bulk Order

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Quality Level

sustainability

Greener Alternative Product

concentration

2.5 wt. %

work function

3.9 eV, variation

viscosity

3.5 cP

greener alternative category

SMILES string

[Al][Zn]=O

InChI

1S/Al.O.Zn

InChI key

PZVKIZCGFVBSGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This aluminium-doped (3.15 mol% Al) zinc oxide nanoparticle ink is a nanoparticle-based printing ink allowing processing temperatures of 80-120 °C. The Al-doped Zinc oxide nanoparticle ink is an electron selective interface layer ink containing Al-doped zinc oxide (AZO) nanoparticles in butanols. AZO nanoparticle ink functions well as electron extraction layer (EEL) materials in solar cells.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is used as an Optical Spacers in Color-Tuned Highly Efficient Organic Solar Cells and has been enhanced for energy efficiency. Click here for more information.

Application

This Al-doped ZnO nanoparticle ink is for slot-die, spin-coating, doctor blading or inkjet printing for the use as electron transport layer in printed electronics. The Al-doped ZnO nanoparticle ink is universally applicable in normal and inverted architecture.

Annealing temperature 80-120°C.

Preparation Note

  • Storage: In dark at room temperature
  • Prior to application: Shake, ultrasonicate and (optionally) filter through 0.45 μm PTFE filter
  • Post-treatment: Annealing of deposited Al-doped ZnO films at 80-120°C
  • Thickness optimization may be necessary

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find advantages of inorganic interface layer inks for organic electronic & other applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service