Skip to Content
Merck
All Photos(1)

Key Documents

792551

Sigma-Aldrich

H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH

98% (HPLC)

Synonym(s):

(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid, Schultz Fluorescent UAA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,870.00

€1,870.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€1,870.00

About This Item

Empirical Formula (Hill Notation):
C13H13NO5
Molecular Weight:
263.25
UNSPSC Code:
12352111
NACRES:
NA.22

€1,870.00


Check Cart for Availability

Request a Bulk Order

Assay

98% (HPLC)

form

solid

optical activity

[α]22/D +8.0°, c = 0.5% in 1 M NaOH

mp

276-281 °C

storage temp.

15-25°C

Application

The courmarin-based unnatural amino acid is an effective fluorophore; which has been incoporated into several studies of protein interactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishu Saraogi et al.
Journal of the American Chemical Society, 133(38), 14936-14939 (2011-08-30)
As newly synthesized proteins emerge from the ribosome, they interact with a variety of cotranslational cellular machineries that facilitate their proper folding, maturation, and localization. These interactions are essential for proper function of the cell, and the ability to study
Douglas D Young et al.
Bioorganic & medicinal chemistry letters, 21(24), 7502-7504 (2011-11-02)
The site-specific incorporation of unnatural amino acids (UAAs) into proteins in bacteria is made possible by the evolution of aminoacyl-tRNA synthetases that selectively recognize and aminoacylate the amino acid of interest. Recently we have discovered that some of the previously
Jiangyun Wang et al.
Journal of the American Chemical Society, 128(27), 8738-8739 (2006-07-06)
The fluorescent amino acid l-(7-hydroxycoumarin-4-yl) ethylglycine 1 has been genetically encoded in E. coli in response to the amber TAG codon. Because of its high fluorescence quantum yield, relatively large Stoke's shift, and sensitivity to both pH and polarity, this
Timo Koopmans et al.
Bioorganic & medicinal chemistry, 21(2), 553-559 (2012-12-15)
Incorporation of the unnatural amino acid L-(7-hydroxycoumarin-4-yl)ethylglycine (7-HC) is a powerful and reliable approach for the preparation of fluorescently labeled proteins. The growing popularity of this valuable amino acid prompted us to pursue an improved protocol for its synthetic preparation.
Shengxi Chen et al.
Journal of the American Chemical Society, 135(35), 12924-12927 (2013-08-15)
Two fluorescent amino acids, including the novel fluorescent species 4-biphenyl-l-phenylalanine (1), have been incorporated at positions 17 and 115 of dihydrofolate reductase (DHFR) to enable a study of conformational changes associated with inhibitor binding. Unlike most studies involving fluorescently labeled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service