Skip to Content
Merck
All Photos(1)

Key Documents

792470

Sigma-Aldrich

Me3(OMe)tBuXPhos

96% (HPLC)

Synonym(s):

2-Di-t-butylphosphino-4-methoxy-3,5,6-trimethyl-2′,4′,6′-tri-i-propylbiphenyl, [~1:1 mixture with regioisomer, 2-Di-t-butylphosphino-5-methoxy-3,4,6-trimethyl-2′,4′,6′-tri-i-propylbiphenyl]

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€101.00
1 G
€274.00
5 G
€1,090.00

€101.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
250 MG
€101.00
1 G
€274.00
5 G
€1,090.00

About This Item

Empirical Formula (Hill Notation):
C33H53OP
Molecular Weight:
496.75
UNSPSC Code:
12350000
NACRES:
NA.22

€101.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

96% (HPLC)

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

125-135 °C

functional group

phosphine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Ueda et al.
The Journal of organic chemistry, 77(5), 2543-2547 (2012-02-09)
A new biarylphosphine ligand, Me(3)(OMe)tBuXPhos (L3), was designed as a surrogate for Me(4)tBuXPhos (L1). The Me(3)(OMe)tBuXPhos could be prepared in a chromatography-free manner from inexpensive and readily available 2,3,6-trimethylphenol. Comparative studies demonstrated that a catalyst based on Me(3)(OMe)tBuXPhos displayed the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service