Skip to Content
Merck
All Photos(6)

Key Documents

730998

Sigma-Aldrich

tBuBrettPhos

97%

Synonym(s):

t-Bu Brett Phos, t-BuBrett-Phos, tertButylBrettPhos, 2-(Di-tert-butylphosphino)-2′,4′,6′- triisopropyl-3,6-dimethoxy-1,1′-biphenyl, t-BuBrett Phos, t-BuBrettPhos, [3,6-Dimethoxy-2′,4′,6′-tris(1-methylethyl) [1,1′-biphenyl]-2-yl]bis(1,1-dimethylethyl)phosphine, tert-ButylBrettPhos

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€141.00
500 MG
€189.00
1 G
€258.00
5 G
€913.00
50 G
€5,340.00

€141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€141.00
500 MG
€189.00
1 G
€258.00
5 G
€913.00
50 G
€5,340.00

About This Item

Empirical Formula (Hill Notation):
C31H49O2P
CAS Number:
Molecular Weight:
484.69
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

€141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

reagent type: ligand
reaction type: Fluorinations

mp

166-170 °C

functional group

phosphine

SMILES string

COc1ccc(OC)c(c1P(C(C)(C)C)C(C)(C)C)-c2c(cc(cc2C(C)C)C(C)C)C(C)C

InChI

1S/C31H49O2P/c1-19(2)22-17-23(20(3)4)27(24(18-22)21(5)6)28-25(32-13)15-16-26(33-14)29(28)34(30(7,8)9)31(10,11)12/h15-21H,1-14H3

InChI key

REWLCYPYZCHYSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tBuBrettPhos is a dialkylbiaryl phosphine ligand developed by the Buchwald group. It promotes cross-coupling reactions more efficiently and exhibits improved reactivity compared to other catalytic systems.

tBuBrettPhos is a phosphine ligand widely used in palladium-catalyzed cross-coupling reactions.[1]

Application

Common applications
  • Buchwald-Hartwig amination and C-O coupling
  • Suzuki, Negishi, Stille, Hiyama, Sonogashira cross-couplings
  • α-Arylation reaction


New Applications:
  • Conversion of aryl and vinyl triflates to bromides and chlorides[2]
  • Conversion of aryl triflates to aryl fluorides[3]
  • O-Arylation of ethyl acetohydroximate[4]
  • Conversion of aryl chlorides and sulfonates to nitroaromatics[5]

Features and Benefits

  • White crystalline solid
  • Air- and moisture-stable
  • Thermally stable
  • Highly efficient
  • Wide functional group tolerance
  • Excellent selectivity and conversion

Legal Information

Usage subject to US Patent 7,858,784

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-Catalyzed Conversion of Aryl Chlorides, Triflates, and Nonaflates to Nitroaromatics.
Fors BP, et al.
Journal of the American Chemical Society, 131, 12898-12898 (2009)
Pd-Catalyzed O-Arylation of Ethyl Acetohydroximate: Synthesis of O-Arylhydroxylamines and Substituted Benzofuran
Maimone TJ, et al.
Journal of the American Chemical Society, 132, 9990-9990 (2010)
Transition-metal-catalyzed cross-couplings through carbene migratory insertion
Xia Y, et al.
Chemical Reviews, 117(23), 13810-13889 (2017)
Formation of ArF from LPdAr (F): Catalytic conversion of aryl triflates to aryl fluorides.
Watson DA, et al.
Science, 325, 1661-1661 (2009)
Xiaoqiang Shen et al.
Journal of the American Chemical Society, 132(40), 14076-14078 (2010-09-23)
The palladium-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) has been developed using dialkylbiaryl phosphine ligands. A variety of aryl, heteroaryl, and vinyl halides can be prepared via this method in good to

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service