Skip to Content
Merck
All Photos(1)

Key Documents

779520

Sigma-Aldrich

1-Azido-2-(trifluoromethyl)benzene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

2-(Trifluoromethyl)phenyl azide solution, o-Azido-α,α,α-trifluorotoluene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3N3
CAS Number:
Molecular Weight:
187.12
Beilstein:
1457457
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

FC(F)(F)c1ccccc1N=[N+]=[N-]

InChI

1S/C7H4F3N3/c8-7(9,10)5-3-1-2-4-6(5)12-13-11/h1-4H

InChI key

KNPDKRVXJOVOQP-UHFFFAOYSA-N

Application

Review[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Bräse et al.
Angewandte Chemie (International ed. in English), 44(33), 5188-5240 (2005-08-16)
Since the discovery of organic azides by Peter Griess more than 140 years ago, numerous syntheses of these energy-rich molecules have been developed. In more recent times in particular, completely new perspectives have been developed for their use in peptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service