Skip to Content
Merck
All Photos(1)

Key Documents

727482

Sigma-Aldrich

1-Azido-4-chlorobenzene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

4-Chlorophenyl azide solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€207.00

€207.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€207.00

About This Item

Empirical Formula (Hill Notation):
C6H4ClN3
CAS Number:
Molecular Weight:
153.57
Beilstein:
1210408
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€207.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C6H4ClN3/c7-5-1-3-6(4-2-5)9-10-8/h1-4H

InChI key

HZVGOEUGZJFTNN-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service