Skip to Content
Merck
All Photos(1)

Key Documents

464872

Sigma-Aldrich

Lithium diphenylphosphide solution

0.5 M in THF

Synonym(s):

Diphenyl lithium phosphide, Diphenylphosphine lithium salt, Diphenylphosphino)lithium, Lithiodiphenyl phosphide, Lithiodiphenylphosphine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€256.00

€256.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€256.00

About This Item

Linear Formula:
(C6H5)2PLi
CAS Number:
Molecular Weight:
192.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€256.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

concentration

0.5 M in THF

bp

65-67 °C

density

0.925 g/mL at 25 °C

SMILES string

[Li]P(c1ccccc1)c2ccccc2

InChI

1S/C12H10P.Li/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;/h1-10H;/q-1;+1

InChI key

WKUYEGHEUWHKIU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service