Skip to Content
Merck
All Photos(1)

Key Documents

777633

Sigma-Aldrich

4-Aminopyridazine

97%

Synonym(s):

4-Pyridazinamine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€57.90

€57.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€57.90

About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

127-133 °C

storage temp.

−20°C

SMILES string

Nc1ccnnc1

InChI

1S/C4H5N3/c5-4-1-2-6-7-3-4/h1-3H,(H2,5,6)

InChI key

LUCGBEPEAUHERV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service