Skip to Content
Merck
All Photos(1)

Key Documents

732257

Sigma-Aldrich

5-Aminopyrimidine

96%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€93.40

€93.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€93.40

About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€93.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

170-174 °C

SMILES string

Nc1cncnc1

InChI

1S/C4H5N3/c5-4-1-6-3-7-2-4/h1-3H,5H2

InChI key

FVLAYJRLBLHIPV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Casey E O'Neill et al.
Psychopharmacology, 231(16), 3179-3188 (2014-02-25)
Adenosine receptor stimulation and blockade have been shown to modulate a variety of cocaine-related behaviors. These studies identify the direct effects of adenosine receptor stimulation on cocaine seeking during extinction training and the persistent effects on subsequent reinstatement to cocaine
Hongyan Li et al.
Visual neuroscience, 31(3), 237-243 (2014-05-23)
Electrical coupling of photoreceptors through gap junctions suppresses voltage noise, routes rod signals into cone pathways, expands the dynamic range of rod photoreceptors in high scotopic and mesopic illumination, and improves detection of contrast and small stimuli. In essentially all

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service