Skip to Content
Merck
All Photos(1)

Key Documents

775525

Sigma-Aldrich

Acridine-9-carboxaldehyde

97%

Synonym(s):

Acridine-9-carbaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€104.00

€104.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€104.00

About This Item

Empirical Formula (Hill Notation):
C14H9NO
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€104.00


Estimated to ship on01 April 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

144-149 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

O=Cc1c2ccccc2nc3ccccc13

InChI

1S/C14H9NO/c16-9-12-10-5-1-3-7-13(10)15-14-8-4-2-6-11(12)14/h1-9H

InChI key

ISOCABSXIKQOOV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S M Furst et al.
International journal of immunopharmacology, 17(5), 445-452 (1995-05-01)
Carbamazepine, a widely used anticonvulsant, is associated with a wide range of adverse reactions including agranulocytosis, aplastic anemia and drug-induced lupus. It has also been reported to alter immune function in a variety of ways. We had previously demonstrated that
Olivier Mathieu et al.
Xenobiotica; the fate of foreign compounds in biological systems, 41(2), 91-100 (2010-11-23)
Carbamazepine (CBZ) is a useful anticonvulsive drug associated with rare severe adverse drug reactions. The physio-pathological mechanisms of these reactions are unknown although evidence of immunological activation has been reported. The ability of 9-acridinecarboxaldehyde, a CBZ metabolite, to interact with
Olivier Mathieu et al.
Pharmacological reports : PR, 63(1), 86-94 (2011-03-29)
Carbamazepine is a widely used anticonvulsive agent. Its metabolic pathway leads not only to the major active metabolite, carbamazepine-10,11-epoxide, but also to minor terminal metabolites such as iminostilbene and acridine. Carbamazepine is usually well-tolerated, but it may lead to rare
Wolf-Ulrich Palm
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 17(7), 964-974 (2018-06-20)
Dark and photolysis reactions in solution were investigated for 9-acridinecarboxaldehyde (ACL). ACL reacts in the dark at T = 20 °C and pH = 7.0 in an air saturated solution to the main product 9-acridinecarboxylic acid (ACA) and to the
Mei Lu et al.
Journal of the American Society for Mass Spectrometry, 26(10), 1676-1685 (2015-08-06)
Electrochemistry (EC) combined with mass spectrometry (MS) is a powerful tool for elucidation of electrochemical reaction mechanisms. However, direct online analysis of electrochemical reaction in aqueous phase was rarely explored. This paper presents the online investigation of several electrochemical reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service