Skip to Content
Merck
All Photos(1)

Documents

774030

Sigma-Aldrich

Zirconium(IV) oxide

sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis (excludes 2% HfO2)

Synonym(s):

Zirconia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis (excludes 2% HfO2)

form

powder

reaction suitability

core: zirconium

diam. × thickness

2.00 in. × 0.25 in.

bp

5000 °C (lit.)

mp

2700 °C (lit.)

density

5.89 g/mL at 25 °C (lit.)

SMILES string

O=[Zr]=O

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid Oxide Fuel cells operating at temperatures below 800 C (also known as intermediate temperature solid oxide fuel cell, IT-SOFC) are currently the topic of much research and development owing to the high degradation rates and materials costs incurred for SOFC operating at temperatures above 900 C. Thin films of electrode and electrolyte layers is one of the ways to achieve high performances in IT-SOFC.
Zirconium oxide sputtering target can be used for physical vapor deposition of zirconia thin films for IT-SOFC, thermal barrier coatings, lead zirconium titanate (PZT) films for sensor applications and other zirconium containing films for different applications.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay Lin et al.
The Journal of general virology, 95(Pt 3), 557-570 (2013-12-04)
A novel virus was detected in a sample collected from a Swedish moose (Alces alces). The virus was suggested as a member of the Hepeviridae family, although it was found to be highly divergent from the known four genotypes (gt1-4)
D Stender et al.
Physical chemistry chemical physics : PCCP, 17(28), 18613-18620 (2015-06-30)
The crystallization kinetics of amorphous 3 and 8 mol% yttria stabilized zirconia (3YSZ and 8YSZ) thin films grown by pulsed laser deposition (PLD), spray pyrolysis and dc-magnetron sputtering are explored. The deposited films were heat treated up to 1000 °C
Alireza Abdolrasouli et al.
mBio, 6(3), e00536-e00536 (2015-06-04)
A rapid and global emergence of azole resistance has been observed in the pathogenic fungus Aspergillus fumigatus over the past decade. The dominant resistance mechanism appears to be of environmental origin and involves mutations in the cyp51A gene, which encodes
Neil M Ferguson et al.
Science translational medicine, 7(279), 279ra37-279ra37 (2015-03-20)
Dengue is the most common arboviral infection of humans and is a public health burden in more than 100 countries. Aedes aegypti mosquitoes stably infected with strains of the intracellular bacterium Wolbachia are resistant to dengue virus (DENV) infection and
Masayuki Kanki et al.
Toxicology, 324, 158-168 (2014-05-28)
Despite increased focus in recent years on urinary microRNA (miRNA) biomarkers in patients with diabetes and chronic kidney diseases, few studies have explored urinary miRNA markers in drug-induced nephrotoxicity. Here, we attempted to identify urinary miRNA markers suitable for use

Articles

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service