Skip to Content
Merck
Zirconium(IV) oxide Technipur®

Zirconium(IV) oxide

Zirconium(IV) oxide Technipur®
Synonym(s):
Zirconia
Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core