Skip to Content
Merck
All Photos(1)

Key Documents

767123

Sigma-Aldrich

Polyester bis-MPA dendron, 16 hydroxyl, 1 thiol

generation 4

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€690.00

€690.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€690.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

€690.00


Estimated to ship on19 May 2025


Request a Bulk Order

description

hydroxyl surface groups

Quality Level

form

solid

mol wt

generation 4

feature

focal point thiol

no. Surface Groups

16

mp

315-340 °C

storage temp.

−20°C

General description

calculated mol. wt. 1819.9

Application

Thiol dendrons can be conjugated to varius noble surfaces or coupled to unsaturated carbon-carbon bonds using various radical initiators.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service