Skip to Content
Merck
All Photos(1)

Key Documents

760544

Sigma-Aldrich

6-Chloro-1H-benzimidazole-2-carboxylic acid monohydrate

97%

Synonym(s):

5-Chloro-1H-benzimidazole-2-carboxylic acid, 5-Chlorobenzimidazole-2-carboxylic acid, 6-Chlorobenzimidazole-2-carboxylic acid, 5-Chloro-1H-benzimidazole-2-carboxylic acid monohydrate, 5-Chlorobenzimidazole-2-carboxylic acid monohydrate, 6-Chlorobenzimidazole-2-carboxylic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5ClN2O2 · H2O
CAS Number:
Molecular Weight:
214.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

156-161 °C

functional group

carboxylic acid
chloro

SMILES string

ClC1=CC2=C(N=C(C(O)=O)N2)C=C1.O

InChI

1S/C8H5ClN2O2.H2O/c9-4-1-2-5-6(3-4)11-7(10-5)8(12)13;/h1-3H,(H,10,11)(H,12,13);1H2

InChI key

GSFFORBRGSPAKD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service