Skip to Content
Merck
All Photos(1)

Key Documents

242098

Sigma-Aldrich

N-Methyltrifluoroacetamide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€103.00

About This Item

Linear Formula:
CF3CONHCH3
CAS Number:
Molecular Weight:
127.07
Beilstein:
1703392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

156-157 °C (lit.)

mp

49-51 °C (lit.)

functional group

amide
fluoro

SMILES string

CNC(=O)C(F)(F)F

InChI

1S/C3H4F3NO/c1-7-2(8)3(4,5)6/h1H3,(H,7,8)

InChI key

IQNHBUQSOSYAJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The high-resolution proton and fluorine resonance spectra of N-methyltrifluoroacetamide in tetrahydrofuran and acetone was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Yoshida
Environmental toxicology and pharmacology, 37(3), 1123-1128 (2014-05-06)
Recently, a pyrethroid profluthrin [(2,3,5,6-tetrafluoro-4-methylphenyl)methyl 2,2-dimethyl-3-(prop-1-enyl)cyclopropane-1-carboxylate] is widely used as mothproof repellents in indoors. The urinary excretion kinetics of its metabolites was examined in rats to search for urinary metabolites suitable as biomarkers of profluthrin exposure in the general population.
Sabrina Lechner et al.
International journal of medical microbiology : IJMM, 304(8), 931-940 (2014-07-02)
Treatment of stationary growth phase Staphylococcus aureus SA113 with 100-fold of the MIC of the lipopeptide antibiotic daptomycin leaves alive a small fraction of drug tolerant albeit genetically susceptible bacteria. This study shows that cells of this subpopulation exhibit active
Xianqin Wang et al.
Biological & pharmaceutical bulletin, 38(3), 470-475 (2015-03-12)
Numerous people die of paraquat (PQ) poisoning every year in the world. Although several studies regarding paraquat (PQ) poisoning have been conducted, the metabolic changes in plasma remain unknown. In this study, the metabolomics of 15 PQ poisoned patients with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service