Skip to Content
Merck
All Photos(3)

Key Documents

760048

Sigma-Aldrich

2-Chloro-4-methoxybenzyl bromide

97%

Synonym(s):

1-(Bromomethyl)-2-chloro-4-methoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€135.00

€135.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€135.00

About This Item

Empirical Formula (Hill Notation):
C8H8BrClO
CAS Number:
Molecular Weight:
235.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€135.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

53-57 °C

functional group

bromo
chloro

storage temp.

−20°C

SMILES string

COc1ccc(CBr)c(Cl)c1

InChI

1S/C8H8BrClO/c1-11-7-3-2-6(5-9)8(10)4-7/h2-4H,5H2,1H3

InChI key

UIOYCWAQKOLSMA-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

219.9 °F

Flash Point(C)

104.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service