Skip to Content
Merck
All Photos(1)

Key Documents

759864

Sigma-Aldrich

1,2-Difluorobenzene-d4

97 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4F2
Molecular Weight:
118.12
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

97 atom % D

Assay

98% (CP)

form

liquid

mass shift

M+4

SMILES string

[2H]C1=C([2H])C([2H])=C([2H])C(F)=C1F

InChI

1S/C6H4F2/c7-5-3-1-2-4-6(5)8/h1-4H/i1D,2D,3D,4D

InChI key

GOYDNIKZWGIXJT-RHQRLBAQSA-N

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
768502790826731021
assay

98% (CP)

assay

≥98% (CP)

assay

98% (CP)

assay

98% (CP)

isotopic purity

97 atom % D

isotopic purity

≥98 atom % D

isotopic purity

99 atom % D

isotopic purity

98 atom % D

form

liquid

form

liquid

form

liquid

form

liquid

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service