Skip to Content
Merck
All Photos(2)

Key Documents

757144

Sigma-Aldrich

Vinylene carbonate

greener alternative

contains 80 ppm BHT as stabilizer, 99%

Synonym(s):

1,3-Dioxol-2-one, VC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2O3
CAS Number:
Molecular Weight:
86.05
Beilstein:
105683
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

80 ppm BHT as stabilizer

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.421 (lit.)
n20/D 1.422

bp

162 °C (lit.)

mp

19-22 °C (lit.)

density

1.355 g/mL at 25 °C (lit.)
1.355 g/mL at 25 °C

application(s)

battery manufacturing

greener alternative category

storage temp.

2-8°C

SMILES string

O=C1OC=CO1

InChI

1S/C3H2O3/c4-3-5-1-2-6-3/h1-2H

InChI key

VAYTZRYEBVHVLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Vinylene carbonate (VC) can be used as an electrolytic additive for ionic liquid electrolyte for the formation of lithium-ion batteries. It can also be used in the formation of hybrid solid electrolyte interphase (SEI) for the fabrication of lithium-based batteries.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Stomach

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

181.9 °F - closed cup

Flash Point(C)

83.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinylene carbonate-LiNO3: a hybrid additive in carbonic ester electrolytes for SEI modification on Li metal anode
Guo J, et al.
Electrochemical Communications, 51(15), 59-63 (2015)
Electrochemical investigations of ionic liquids with vinylene carbonate for applications in rechargeable lithium ion batteries
Sun X and Dai S
Electrochimica Acta, 55(15), 4618-4626 (2010)
Matthieu Groh et al.
European journal of internal medicine, 26(7), 545-553 (2015-05-15)
To develop disease-specific recommendations for the diagnosis and management of eosinophilic granulomatosis with polyangiitis (Churg-Strauss syndrome) (EGPA). The EGPA Consensus Task Force experts comprised 8 pulmonologists, 6 internists, 4 rheumatologists, 3 nephrologists, 1 pathologist and 1 allergist from 5 European
Surface analytical study regarding the solid electrolyte interphase composition of nanoparticulate SnO2 anodes for Li-ion batteries
Winkler V, et al.
The Journal of Physical Chemistry C, 120(43), 24706-24714 (2016)
Stefan Hilf et al.
Journal of the American Chemical Society, 130(33), 11040-11048 (2008-07-24)
The selective placement of a functional group at the chain end of a ring-opening metathesis polymer using ruthenium carbene initiators has been a significant limitation. Here we demonstrate a highly effective and facile end-capping technique for ROMP with living ruthenium

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Solid oxide fuel cells and electrolyzers show potential for chemical-to-electrical energy conversion, despite early development stages.

Li-ion batteries are currently the focus of numerous research efforts with applications designed to reduce carbon-based emissions and improve energy storage capabilities.

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service