Skip to Content
Merck
All Photos(1)

Key Documents

757063

Sigma-Aldrich

(2R,3S)-Dimethyl oxirane-2,3-dicarboxylate

97%

Synonym(s):

Dimethyl cis-2,3-epoxysuccinate, Dimethyl cis-oxirane-2,3-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€64.50

€64.50


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€64.50

About This Item

Empirical Formula (Hill Notation):
C6H8O5
CAS Number:
Molecular Weight:
160.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.50


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.440

density

1.269 g/mL at 25 °C

functional group

ester
ether

SMILES string

COC(=O)[C@H]1O[C@H]1C(=O)OC

InChI

1S/C6H8O5/c1-9-5(7)3-4(11-3)6(8)10-2/h3-4H,1-2H3/t3-,4+

InChI key

KAGHUYFLGPUYKE-ZXZARUISSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service