Skip to Content
Merck
All Photos(1)

Key Documents

43200

Sigma-Aldrich

trans-1,3-Diphenyl-2-propen-1-ol

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€118.00

€118.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€118.00

About This Item

Empirical Formula (Hill Notation):
C15H14O
CAS Number:
Molecular Weight:
210.27
Beilstein:
1951697
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€118.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

solid

mp

55-57 °C

functional group

hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

OC(\C=C\c1ccccc1)c2ccccc2

InChI

1S/C15H14O/c16-15(14-9-5-2-6-10-14)12-11-13-7-3-1-4-8-13/h1-12,15-16H/b12-11+

InChI key

ORACYDGVNJGDMI-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-1,3-diphenyl-2-propen-1-ol is an allylic alcohol. It has been reported to exhibit significant in vivo anti-inflammatory activity.[1] Allylic amination of trans-1,3-diphenyl-2-propen-1-ol catalyzed by water-soluble calix[4]resorcinarene sulfonic acid has been reported.[2]

Application

trans-1,3-diphenyl-2-propen-1-ol may be used as a model substrate to investigate the formation of substituted cyclopropanes by SiO2-ZrO2 mixed oxides catalyzed Friedel-Crafts-alkylation followed by trans-hydrogenation.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Nucleophilic SN1-Type Reactions of Alcohols.
Emer E, et al.
European Journal of Organic Chemistry, 4, 647-666 (2011)
Xinqin Liu et al.
International journal of biological sciences, 13(11), 1351-1360 (2017-12-07)
Lead (Pb) is a well-known neurotoxicant and a risk factor for neurologic disorders. The blood brain barrier (BBB) plays an important role in the maintenance of optimal brain function. BBB is a target of Pb, and studies have shown that
Molecular topology as a novel approach for drug discovery.
Galvez J, et al.
Expert Opinion on Drug Delivery, 7(2), 133-153 (2012)
High-Surface-Area SiO2-ZrO2 Mixed Oxides as Catalysts for the Friedel-Crafts-Type Alkylation of Arenes with Alcohols and Tandem Cyclopropanation Reactions.
Kaper H, et al.
ChemCatChem, 4(11), 1813-1818 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service