Skip to Content
Merck
All Photos(1)

Key Documents

752940

Sigma-Aldrich

(Trimethylphosphoranylidene)acetonitrile solution

0.5 M in THF

Synonym(s):

CMMP, Cyanomethylenetrimethylphosphorane solution

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€274.00
10 ML
€489.00

€274.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€274.00
10 ML
€489.00

About This Item

Empirical Formula (Hill Notation):
C5H10NP
CAS Number:
Molecular Weight:
115.11
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

€274.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

0.5 M in THF

density

0.896 g/mL at 25 °C

functional group

nitrile
phosphine

storage temp.

2-8°C

SMILES string

CP(C)(C)=CC#N

InChI

1S/C5H10NP/c1-7(2,3)5-4-6/h5H,1-3H3

InChI key

KJFJIEHJCVSAKJ-UHFFFAOYSA-N

General description

(Trimethylphosphoranylidene)acetonitrile is a reagent generally employed in the Wittig olefination of less reactive carboxylic acid compounds like esters, N-Boc lactams, lactones, and imides to give α,β-unsaturated nitriles. It is also used as an alternate reagent for Mitsonobu reaction by replacing DEAD/PPh3 for the alkylation of C-H and N-H bonds.[1][2]

Other Notes

If precipitate is observed, warm gently to resolubilize

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-5.8 °F - closed cup

Flash Point(C)

-21.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyanomethylenetrimethylphosphorane
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2003)
Cyanomethylenetrimethylphosphorane
e-EROS Encyclopedia of Reagents for Organic Synthesis (2003)
Cyanomethylenetrimethylphosphorane, a powerful reagent for the Wittig olefination of esters, lactones and imides
Tsunoda T, et al.
Tetrahedron Letters, 41(2), 235-237 (2000)
Junnan Tang et al.
Nature communications, 8, 13724-13724 (2017-01-04)
Stem cell therapy represents a promising strategy in regenerative medicine. However, cells need to be carefully preserved and processed before usage. In addition, cell transplantation carries immunogenicity and/or tumourigenicity risks. Mounting lines of evidence indicate that stem cells exert their
Siomara Hernandez et al.
Neuropharmacology, 179, 108264-108264 (2020-08-08)
Gulf War Illness (GWI) is a chronic multisymptom illness that includes gastrointestinal disorders. Although the exact etiology of GWI is unknown, exposure to the drug pyridostigmine bromide (PB) is considered a major factor. Exposure to PB drives enteric neuroinflammation, promotes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service