Skip to Content
Merck
All Photos(1)

Key Documents

752398

Sigma-Aldrich

Poly(ethylene glycol), 4 hydroxyl dendron, generation 1

average Mn 20,300

Synonym(s):

PFD-G1-PEG20k-OH, Dendron functionalized PEG, generation 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

form

solid

mol wt

PEG average Mn 20,000
average Mn 20,300
generation 1

no. Surface Groups

4

transition temp

Tm 54-58 °C

Mw/Mn

≤1.2

functional group

hydroxyl

Looking for similar products? Visit Product Comparison Guide

Other Notes

average PEG Mn = 20,000; esterified with 4 OH groups

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qigang Wang et al.
Nature, 463(7279), 339-343 (2010-01-22)
With the world's focus on reducing our dependency on fossil-fuel energy, the scientific community can investigate new plastic materials that are much less dependent on petroleum than are conventional plastics. Given increasing environmental issues, the idea of replacing plastics with

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service