Skip to Content
Merck
All Photos(2)

Key Documents

ALD00386

Sigma-Aldrich

Fe(dibm)3

greener alternative

Synonym(s):

Baran Olefin Cross-Coupling Catalyst, Iron tris(diisobutyrylmethane)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€54.00

€54.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€54.00

About This Item

Empirical Formula (Hill Notation):
C27H45FeO6
CAS Number:
Molecular Weight:
521.49
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€54.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

96-102 °C

greener alternative category

SMILES string

O=C([CH-]C(C(C)C)=O)C(C)C.O=C([CH-]C(C(C)C)=O)C(C)C.O=C([CH-]C(C(C)C)=O)C(C)C.[Fe+3]

InChI

1S/3C9H15O2.Fe/c3*1-6(2)8(10)5-9(11)7(3)4;/h3*5-7H,1-4H3;/q3*-1;+3

InChI key

LKNJSJQVZKKQHU-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Iron tris(diisobutyrylmethane) was reported by Baran and coworkers to be an effective catalyst in the cross-coupling of olefins. In the presence of phenylsilane (Aldrich 335150), several highly substituted and novel functionalized C-C bonds can be formed using simple and mild reaction conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service