Skip to Content
Merck
All Photos(1)

Key Documents

745693

Sigma-Aldrich

3-(Aminomethyl)oxetane

97%

Synonym(s):

3-oxetanemethanamine, (Oxetan-3-yl)methanamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€290.00

€290.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€290.00

About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€290.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.458

density

1.014 g/mL at 25 °C

functional group

amine
ether

storage temp.

−20°C

SMILES string

NCC1COC1

InChI

1S/C4H9NO/c5-1-4-2-6-3-4/h4H,1-3,5H2

InChI key

KTHZBRAXOLUNBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

144.9 °F

Flash Point(C)

62.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service