Skip to Content
Merck
All Photos(1)

Key Documents

744743

Sigma-Aldrich

(2-Bromophenyl)(2,4,6-trimethylphenyl)iodonium triflate

≥98% (HPLC)

Synonym(s):

(2-Bromophenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate, 2-Bromophenyl)mesityliodonium triflate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€152.00

€152.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€152.00

About This Item

Empirical Formula (Hill Notation):
C16H15BrF3IO3S
CAS Number:
Molecular Weight:
551.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€152.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

composition

carbon, 34.0-35.7% CH

reaction suitability

reagent type: oxidant

functional group

bromo
fluoro
iodo
triflate

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1cc(C)c([I+]c2ccccc2Br)c(C)c1

InChI

1S/C15H15BrI.CHF3O3S/c1-10-8-11(2)15(12(3)9-10)17-14-7-5-4-6-13(14)16;2-1(3,4)8(5,6)7/h4-9H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

RYZOZGQFOKVCDZ-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service