Skip to Content
Merck
All Photos(1)

Key Documents

671584

Sigma-Aldrich

Di(1-adamantyl)-n-butylphosphine hydriodide

95%

Synonym(s):

Di(1-adamantanyl)-n-butyl-phosphonium iodide, cataCXium® AHI

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€210.00

€210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€210.00

About This Item

Empirical Formula (Hill Notation):
C24H40IP
CAS Number:
Molecular Weight:
486.45
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€210.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: Asymmetric synthesis
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

SMILES string

I.CCCCP([C@]12C[C@H]3C[C@H](C[C@H](C3)C1)C2)[C@@]45C[C@@H]6C[C@@H](C[C@@H](C6)C4)C5

InChI

1S/C24H39P.HI/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24;/h17-22H,2-16H2,1H3;1H/t17-,18+,19-,20-,21+,22-,23-,24-;

InChI key

IBXHWLZKSOGUFS-HDWJELNESA-N

General description

sold in collaboration with Solvias AG

Application

Phosphonium salts are ligand precursors for the palladium-catalyzed amination, and Suzuki coupling

Legal Information

US7148176
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service