Skip to Content
Merck
All Photos(2)

Key Documents

740136

Sigma-Aldrich

4,7-Dihydroxycoumarin

97%

Synonym(s):

4,7-Dihydroxychromen-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€159.00

€159.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€159.00

About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€159.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

97%

form

solid

mp

275-280 °C

functional group

ester
ketone

storage temp.

2-8°C

SMILES string

Oc1ccc2C(O)=CC(=O)Oc2c1

InChI

1S/C9H6O4/c10-5-1-2-6-7(11)4-9(12)13-8(6)3-5/h1-4,10-11H

InChI key

CYSRKZFPSNZSCS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ilkay Erdogan Orhan et al.
Bioorganic chemistry, 84, 355-362 (2018-12-12)
Coumarins of synthetic or natural origins are an important chemical class exerting diverse pharmacological activities. In the present study, 26 novel O-alkylcoumarin derivatives were synthesized and have been tested at 100 µM for their in vitro inhibitory potential against acetylcholinesterase (AChE)
C Preston Pugh et al.
Archives of biochemistry and biophysics, 564, 244-253 (2014-12-03)
The widely used anticoagulant Coumadin (R/S-warfarin) undergoes oxidation by cytochromes P450 into hydroxywarfarins that subsequently become conjugated for excretion in urine. Hydroxywarfarins may modulate warfarin metabolism transcriptionally or through direct inhibition of cytochromes P450 and thus, UGT action toward hydroxywarfarin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service