Skip to Content
Merck
All Photos(1)

Key Documents

382272

Sigma-Aldrich

Chelidonic acid

≥95%

Synonym(s):

4-Oxo-4H-pyran-2,6-dicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€163.00
25 G
€590.00

€163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€163.00
25 G
€590.00

About This Item

Empirical Formula (Hill Notation):
C7H4O6
CAS Number:
Molecular Weight:
184.10
Beilstein:
163607
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

265 °C (dec.) (lit.)

functional group

carboxylic acid
ether
ketone

SMILES string

OC(=O)C1=CC(=O)C=C(O1)C(O)=O

InChI

1S/C7H4O6/c8-3-1-4(6(9)10)13-5(2-3)7(11)12/h1-2H,(H,9,10)(H,11,12)

InChI key

PBAYDYUZOSNJGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chelidonic acid (CA) is a γ-pyrone.[1] It is reported as constituent of the rhizome of Chelidonium majus L. It has many pharmacological effects, such as mild analgesic and antimicrobial effects.[2] Therapeutic potential of CA for the treatment of intestinal inflammation has been investigated.[2] CA is reported as inhibitor of the rat brain glutamate decarboxylase.[3] Biosynthesis of CA in cell suspension cultures of Leucojum aestivum has been studied.[4]

Application

Chelidonic acid is suitable for use a in homogeneous preparation of native dihydrodipicolinate synthase from pea.[5] It may be used as endocyclic oxygen-containing ligand in the synthesis of aqua[bis(2-pyridylmethyl)amine][chelidonato(1.5-)]-copper(II) chelidonate(0.5-) monohydrate.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Dereppe et al.
Plant physiology, 98(3), 813-821 (1992-03-01)
Dihydrodipicolinate synthase (EC 4.2.1.52), the first enzyme unique to lysine biosynthesis in bacteria and higher plants, has been purified to homogeneity from etiolated pea (Pisum sativum) seedlings using a combination of conventional and affinity chromatographic steps. This is the first
Aqua [bis (2-pyridylmethyl) amine][chelidonato (1.5-)] copper (II) chelidonate (0.5-) monohydrate.
Fainerman-Melnikova M, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(12), m3582-m3584 (2006)
Mohammad Ghadermazi et al.
Acta crystallographica. Section C, Crystal structure communications, 67(Pt 4), o134-o138 (2011-04-07)
Two related proton-transfer compounds, namely piperazine-1,4-diium 4-oxo-4H-pyran-2,6-dicarboxylate monohydrate, C(4)H(12)N(2)(2+)·C(7)H(2)O(6)(2-)·H(2)O or (pipzH(2))(cdo)·H(2)O, (I), and piperazine-1,4-diium bis(6-carboxy-4-oxo-4H-pyran-2-carboxylate), C(4)H(12)N(2)(2+)·2C(7)H(3)O(6)(-) or (pipzH(2))(cdoH)(2), (II), were obtained by the reaction of 4-oxo-4H-pyran-2,6-dicarboxylic acid (chelidonic acid, cdoH(2)) and piperazine (pipz). In (I), both carboxyl H atoms of
Shen Z-W et al.
Phytochemistry, 57(1), 33-42 (2001-05-05)
The biosynthesis of chelidonic acid was studied in cell suspension cultures of Leucojum aestivum. Cell cultures were supplied with [U-13C]glucose, [l-13C]glucose or [U-13Cs]ribose/ribulose in standard medium containing unlabeled glucose. 13C labeling patterns of amino acids obtained by hydrolysis of biomass
Dae-Seung Kim et al.
Biological & pharmaceutical bulletin, 35(5), 666-671 (2012-06-13)
Chelidonic acid (CA), a constituent of Chelidonium majus L., has many pharmacological effects, including mild analgesic and antimicrobial effects. However, the effects of CA on intestinal inflammation and the molecular mechanisms responsible are poorly understood. The aim of this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service